Anti-TYRP1 Mouse Monoclonal Antibody [clone: TA99 TYRP1/807]

Supplier: Biotium

BNUB0808-100 BNUB0808-500
75977-906EA 380.65 CAD
75977-906 75977-908
Anti-TYRP1 Mouse Monoclonal Antibody [clone: TA99 TYRP1/807]
Antibodies
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: Clone TA99 TYRP1/807
Conjugation:More Product Information
Order Now

SPECIFICATIONS

Antigen symbol TYRP1
Antigen name TRP1
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone TA99 TYRP1/807
Reactivity Human,Mouse
Host Mouse
Gene ID Unigene ID: 270279
Isotype IgGs
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.2 mg/mL
Storage buffer PBS/0.05% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR