Anti-TYRP1 Mouse Monoclonal Antibody (CF594) [clone: TYRP1/807]

Supplier: Biotium

BNC940807-100 BNC940807-500
75978-044EA 325.12 CAD
75978-044 75978-046
Anti-TYRP1 Mouse Monoclonal Antibody (CF594) [clone: TYRP1/807]
Antibodies
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: Clone TYRP1/807
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR