Anti-TYRP1 Mouse Monoclonal Antibody (Biotin) [clone: TA99]

Supplier: Biotium

BNCB0806-100 BNCB0806-500
75977-840EA 325.12 CAD
75977-840 75977-842
Anti-TYRP1 Mouse Monoclonal Antibody (Biotin) [clone: TA99]
Antibodies
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: Clone TA99
Conjugation: Biotin
Epitope:
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol TYRP1
Antigen name TRP1
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone TA99
Reactivity Human,Mouse
Host Mouse
Gene ID Unigene ID: 270279
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR