Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: KRT19/799]

Supplier: Biotium

BNCB0799-100 BNCB0799-500
75922-518EA 325.12 CAD
75922-518 75922-520
Anti-KRT19 Mouse Monoclonal Antibody (Biotin) [clone: KRT19/799]
Antibodies
This antibody recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK1More Product Information
Order Now

SPECIFICATIONS

Antigen symbol KRT19
Environmentally Preferable
Antigen name Cytokeratin 19
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone KRT19/799
Reactivity Human
Host Mouse
Gene ID Unigene ID: 654568
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR