Anti-KRT19 Mouse Monoclonal Antibody (CF488A) [clone: KRT19/799]

Supplier: Biotium

BNC880799-100 BNC880799-500
75919-744EA 357.68 CAD
75919-744 75919-746
Anti-KRT19 Mouse Monoclonal Antibody (CF488A) [clone: KRT19/799]
Antibodies
This antibody recognizes a protein of 40 kDa, identified as cytokeratin-19 (CK19), which is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: Clone KRT19/799
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR