Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF405S) [clone: PY793]

Supplier: Biotium

BNC040793-100 BNC040793-500
75970-152EA 325.12 CAD
75970-152 75970-154
Anti-Phosphotyrosine Mouse Monoclonal Antibody (CF405S) [clone: PY793]
Antibodies
Protein phosphorylation is a fundamental event in the regulation of a large number of intracellular processes. Phosphorylation of specific tyrosine residues is the result of activation or stimulation of their respective protein tyrosine kinases. The phosphorylated proteins can be auto-phosphorylated kinases or certain cellular protein substrates. Tyrosine-phosphorylated proteins are involved in signal transduction and in the regulation of cell proliferation. Antibody to phosphotyrosine provides an excellent tool for the detection, characterization, and purification of phosphotyrosine containing proteins. This MAb shows no cross-reaction with other phosphoamino acids and is superb for multiple applications including staining of formalin/paraffin tissues.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: Clone PY793
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: All species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR