Anti-ECM1 Mouse Monoclonal Antibody (CF405S) [clone: ECM1/792]

Supplier: Biotium

BNC040792-100 BNC040792-500
75952-208EA 325.12 CAD
75952-208 75952-210
Anti-ECM1 Mouse Monoclonal Antibody (CF405S) [clone: ECM1/792]
Antibodies
This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiationMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ECM1
Environmentally Preferable
Antigen name IgA Secretory Component
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 405S
Clone ECM1/792
Reactivity Human,Rat
Host Mouse
Gene ID Unigene ID: 81071
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR