Anti-ACTA2 Mouse Monoclonal Antibody (Biotin) [clone: ACTA2/791]

Supplier: Biotium

BNCB0791-100 BNCB0791-500
75973-132EA 325.12 CAD
75973-132 75973-134
Anti-ACTA2 Mouse Monoclonal Antibody (Biotin) [clone: ACTA2/791]
Antibodies
Actin is a major component of the cytoskeleton and is present in most cell types. It is highly specific to actin from smooth muscles. This MAb does not stain cardiac or skeletal muscle; however, it does stain myofibroblasts and myoepithelial cells. This antibody could be used together with anti-muscle specific actin and myogenin in making a diagnosis of smooth muscle and skeletal muscle tumors. In most cases of rhabdomyosarcoma, this antibody yields negative results whereas anti-muscle specific actin and myogenin are positive. Leiomyosarcomas are positive only with anti-muscle specific actinMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ACTA2
Antigen name Smooth Muscle Actin
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone ACTA2/791
Reactivity Human,Rat
Host Mouse
Gene ID Unigene ID: 500483
Isotype IgG2a kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR