Anti-HSPB1 Mouse Monoclonal Antibody (CF640R) [clone: HSPB1/774]

Supplier: Biotium

BNC400774-100 BNC400774-500
75953-326EA 325.12 CAD
75953-326 75953-328
Anti-HSPB1 Mouse Monoclonal Antibody (CF640R) [clone: HSPB1/774]
Antibodies
This antibody recognizes a 24-27 kDa estrogen-regulated protein, identified as heat shock protein 27 (hsp27). Hsp27 was recently found to be identical to the estrogen-induced p29 and 24K protein. About 50% of breast carcinomas are positive for hsp27 especially those that are also positive for estrogen and/or progesterone receptor. HSP27 has also been implicated in drug resistance in cancer cells.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: HSPB1
Clonality: Monoclonal
Clone: HSPB1/774
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chicken, Chimpanzee, Human, Mouse, Rat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR