Anti-CDKN1B Mouse Monoclonal Antibody [clone: DCS-72.F6]

Supplier: Biotium

BNUB0771-100 BNUB0771-500
75968-962EA 380.65 CAD
75968-962 75968-964
Anti-CDKN1B Mouse Monoclonal Antibody [clone: DCS-72.F6]
Antibodies
This antibody recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR