Anti-CDKN1B Mouse Monoclonal Antibody (Biotin) [clone: KIP1/769]

Supplier: Biotium

BNCB0769-100 BNCB0769-500
75967-320EA 325.12 CAD
75967-320 75967-322
Anti-CDKN1B Mouse Monoclonal Antibody (Biotin) [clone: KIP1/769]
Antibodies
This MAb recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb is excellent for staining of formalin-fixed tissues.

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: Clone KIP1/769
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity:More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CDKN1B
Environmentally Preferable
Antigen name p27 / KIP1
Antibody type Primary
Clonality Monoclonal
Conjugation Biotin
Clone KIP1/769
Reactivity Monkey,Human,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 238990
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR