Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26 BA17]

Supplier: Biotium

BNUB0753-100 BNUB0753-500
75922-448EA 380.65 CAD
75922-448 75922-450
Anti-KRT19 Mouse Monoclonal Antibody [clone: A53-B/A2.26 BA17]
Antibodies
This MAb reacts with the rod domain of human cytokeratin-19 (CK19), a polypeptide of 40 kDa. CK19 is expressed in sweat gland, mammary gland ductal and secretory cells, bile ducts, gastrointestinal tract, bladder urothelium, oral epithelia, esophagus, and ectocervical epithelium. Anti-CK19 reacts with a wide variety of epithelial malignancies including adenocarcinomas of the colon, stomach, pancreas, biliary tract, liver, and breast. Perhaps the most useful application is the identification of thyroid carcinoma of the papillary type, although 50%-60% of follicular carcinomas are also labeled. Anti-CK19 is a useful marker for detection of tumor cells in lymph nodes, peripheral blood, bone marrow and breast cancer.

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: Clone A53-B/A2.26 BA17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG's
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR