Anti-TAG-72 Mouse Monoclonal Antibody (CF488A) [clone: B72.3 CC49]

Supplier: Biotium

BNC880738-100 BNC880738-500
75974-396EA 325.12 CAD
75974-396 75974-398
Anti-TAG-72 Mouse Monoclonal Antibody (CF488A) [clone: B72.3 CC49]
Antibodies
This antibody recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: TAG-72
Clonality: Monoclonal
Clone: Clone B72.3 CC49
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cow, Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR