Anti-TAG-72 Mouse Monoclonal Antibody (CF568) [clone: B72.3 CC49]
Supplier: Biotium
BNC680738-100 BNC680738-500
75975-888EA 325.12 CAD
75975-888 75975-890
Anti-TAG-72 Mouse Monoclonal Antibody (CF568) [clone: B72.3 CC49]
Antibodies
This antibody recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibodyMore Product Information