Anti-TAG-72 Mouse Monoclonal Antibody (CF568) [clone: B72.3 CA72/733]

Supplier: Biotium

BNC680734-100 BNC680734-500
75975-868EA 325.12 CAD
75975-868 75975-870
Anti-TAG-72 Mouse Monoclonal Antibody (CF568) [clone: B72.3 CA72/733]
Antibodies
This antibody recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: TAG-72
Clonality: Monoclonal
Clone: Clone B72.3 CA72/733
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cow, Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR