Anti-TAG-72 Mouse Monoclonal Antibody (Biotin) [clone: CA72/733]

Supplier: Biotium

BNCB0733-100 BNCB0733-500
75974-800EA 325.12 CAD
75974-800 75974-802
Anti-TAG-72 Mouse Monoclonal Antibody (Biotin) [clone: CA72/733]
Antibodies
This antibody recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma.

Type: Primary
Antigen: TAG-72
Clonality: Monoclonal
Clone: Clone CA72/733
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Cow, Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR