Anti-AMACR Rabbit Monoclonal Antibody (CF594) [clone: Rabbit PAb]

Supplier: Biotium

BNC940731-100 BNC940731-500
75979-820EA 325.12 CAD
75979-820 75979-822
Anti-AMACR Rabbit Monoclonal Antibody (CF594) [clone: Rabbit PAb]
Antibodies
This antibody recognizes a protein of 54 kDa, which is identified as AMACR, also known as p504S. It is an enzyme that is involved in bile acid biosynthesis and beta-oxidation of branched-chain fatty acids. AMACR is essential in lipid metabolism. It is expressed in cells of premalignant high-grade prostatic intraepithelial neoplasia (HGPIN) and prostate adenocarcinoma. The majority of the carcinoma cells show a distinct granular cytoplasmic staining reaction. AMACR is present at low or undetectable levels in glandular epithelial cells of normal prostate and benign prostatic hyperplasia. AMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol AMACR
Antigen name AMACR / p504S
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Clone Rabbit PAb
Reactivity Human
Host Rabbit
Gene ID Unigene ID: 508343
Isotype IgG1 kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR