Anti-TP53 Mouse Monoclonal Antibody (CF568) [clone: BP53-12 DO-7]

Supplier: Biotium

BNC680723-100 BNC680723-500
75969-108EA 325.12 CAD
75969-108 75969-110
Anti-TP53 Mouse Monoclonal Antibody (CF568) [clone: BP53-12 DO-7]
Antibodies
This antibody recognizes a 53 kDa protein, which is identified as p53 suppressor gene product. It reacts with the mutant as well as the wild form of p53 under denaturing and non-denaturing conditions. Its epitope maps within the N-terminus (aa 20-25) of p53 oncoprotein. p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to MDM2, SV40 T antigen and human papilloma virus E6 protein. Positive nuclear staining with p53 antibody has been reported to be a negative prognostic factor in breast carcinoma, lung carcinoma, colorectal, and urothelial carcinoma. Anti-p53 positivity has also been used to differentiate uterine serous carcinoma from endometrioid carcinoma as well as to detect intratubular germ cell neoplasia. Mutations involving p53 are found in a wide variety of malignant tumors, including breast, ovarian, bladder, colon, lung, and melanoma.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: Clone BP53-12 DO-7
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cow, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR