Anti-Mitochondrial Marker Mouse Monoclonal Antibody (Biotin) [clone: MTC719]

Supplier: Biotium

BNCB0719-100 BNCB0719-500
75961-538EA 325.12 CAD
75961-538 75961-540
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (Biotin) [clone: MTC719]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals, but not insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells and may be used to stain mitochondria of cells in fixed or frozen tissue sections. It can also be used with paraformaldehyde fixed frozen tissue or cell preparations.

Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: Clone MTC719
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR