Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF568) [clone: MTC719]

Supplier: Biotium

BNC680719-100 BNC680719-500
75961-858EA 325.12 CAD
75961-858 75961-860
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF568) [clone: MTC719]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in cells from a wide variety of animals, but not insects and bacteria. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-More Product Information
Order Now

SPECIFICATIONS

Antigen name Mitochondrial Marker
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone MTC719
Reactivity Human
Host Mouse
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR