Anti-MYOD1 Mouse Monoclonal Antibody (CF594) [clone: 5.8A MYD712]

Supplier: Biotium

BNC940713-500 BNC940713-100
75964-770EA 900.17 CAD
75964-770 75968-360
Anti-MYOD1 Mouse Monoclonal Antibody (CF594) [clone: 5.8A MYD712]
Antibodies
This antibody recognizes a phosphor-protein of 45 kDa, identified as MyoD1. This MAb does not cross react with myogenin, Myf5, or Myf6. Antibody to MyoD1 labels the nuclei of myoblasts in developing muscle tissues. MyoD1 is not detected in normal adult tissue, but is highly expressed in the tumor cell nuclei of rhabdomyosarcomas. Occasionally nuclear expression of MyoD1 is seen in ectomesenchymoma and a subset of Wilm s tumors. Weak cytoplasmic staining is observed in several non-muscle tissues, including glandular epithelium and also in rhabdomyosarcomas, neuroblastomas, Ewing s sarcomas and alveolar soft part sarcomas.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: MYOD1
Clonality: Monoclonal
Clone: Clone 5.8A MYD712
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Chicken, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR