Anti-CHGA Mouse Monoclonal Antibody (CF488A) [clone: LK2H10 PHE5]

Supplier: Biotium

BNC880698-100 BNC880698-500
75910-246EA 325.12 CAD
75910-246 75910-248
Anti-CHGA Mouse Monoclonal Antibody (CF488A) [clone: LK2H10 PHE5]
Antibodies
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raise the possibility of paraganglioma. The co-expression of chromogranin and NSE is typical of neuroendocrine neoplasms. Most pituitary adenomas and prolactinomas readily express chromogranin.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®488A is a green fluorescent dye (Ex/Em 490/515 nm) with excellent brightness and photostability. The dye is minimally charged for less non-specific binding. CF®488A also is compatible with super-resolution imaging by TIRF.

Type: Primary
Antigen: CHGA
Clonality: Monoclonal
Clone: Clone LK2H10 PHE5
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR