Anti-IPO-38 Mouse Monoclonal Antibody [clone: IPO-38]

Supplier: Biotium

BNUB0697-100 BNUB0697-500
75955-206EA 380.65 CAD
75955-206 75955-208
Anti-IPO-38 Mouse Monoclonal Antibody [clone: IPO-38]
Antibodies
This antibody recognizes a protein of 14-16 kDa, which is a novel nuclear antigen of proliferating cells. IPO-38 antigen is present in the nuclei of proliferating cells such as Hodgkins disease and non-Hodgkins lymphomas, different forms of leukemias, breast and colorectal carcinomas, and PHA-stimulated lymphocytes. It is not expressed in the cells of non-stimulated lymphocytes and granulocytes. IPO-38 may be a useful marker of cell proliferation during monitoring of tumor progression.

Type: Primary
Antigen: IPO-38
Clonality: Monoclonal
Clone: Clone IPO-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR