Anti-KRT10 Mouse Monoclonal Antibody [clone: DE-K13]

Supplier: Biotium

BNUB0696-100 BNUB0696-500
75916-150EA 380.65 CAD
75916-150 75916-152
Anti-KRT10 Mouse Monoclonal Antibody [clone: DE-K13]
Antibodies
This antibody recognizes cytokeratin 10 (56.5 kDa) and cytokeratin 13 (53 kDa) in Western blotting. It recognizes only cytokeratin 13 in formalin-fixed, paraffin-embedded tissue sections. It does not react with cytokeratin 10 positive, cytokeratin 13 negative epithelia such as epidermis. However, on frozen sections this MAb serves as differentiation-related marker of all stratified epithelia; it stains all suprabasal cells in both cornifying and non-cornifying stratified epithelia and more differentiated cells of squamous carcinomas.

Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: Clone DE-K13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR