Anti-KRT10 Mouse Monoclonal Antibody (CF568) [clone: DE-K13]
Supplier: Biotium
BNC680696-100 BNC680696-500
75916-168EA 325.12 CAD
75916-168 75916-170
Anti-KRT10 Mouse Monoclonal Antibody (CF568) [clone: DE-K13]
Antibodies
This antibody recognizes cytokeratin 10 (56.5 kDa) and cytokeratin 13 (53 kDa) in Western blotting. It recognizes only cytokeratin 13 in formalin-fixed, paraffin-embedded tissue sections. It does not react with cytokeratin 10 positive, cytokeratin 13 negative epithelia such as epidermis. However, on frozen sections this MAb serves as differentiation-related marker of all stratified epithelia; it stains all suprabasal cells in both cornifying and non-cornifying stratified epithelia and more differentiated cells of squamous carcinomas.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®More Product Information