Anti-PCNA Mouse Monoclonal Antibody (CF640R) [clone: PCNA/694]

Supplier: Biotium

BNC400694-100 BNC400694-500
75969-616EA 325.12 CAD
75969-616 75969-618
Anti-PCNA Mouse Monoclonal Antibody (CF640R) [clone: PCNA/694]
Antibodies
This antibody recognizes a non-histone protein of 36 kDa, which is identified as proliferating cell nuclear antigen (PCNA). It is also known as cyclin or polymerase delta auxiliary protein. Elevated expression of PCNA/cyclin has been shown in the nucleus during late G1 phase immediately before the onset of DNA synthesis, becoming maximal during S-phase and declining during G2 and M phases. This MAb is excellent for multiple applications.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: PCNA
Clonality: Monoclonal
Clone: Clone PCNA/694
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR