Anti-FLT1 Mouse Monoclonal Antibody (CF594) [clone: FLT1/658]

Supplier: Biotium

BNC940658-100 BNC940658-500
75978-900EA 325.12 CAD
75978-900 75978-902
Anti-FLT1 Mouse Monoclonal Antibody (CF594) [clone: FLT1/658]
Antibodies
Three cell membrane receptor tyrosine kinases, Flt-1 (also designated VEGF-R1), Flk-1 (also designated VEGF-R2) and Flt-4, putatively involved in the growth of endothelial cells, are characterized by the presence of seven immunoglobulin-like sequences in their extracellular domain. These receptors exhibit high degrees of sequence relatedness to each other as well as lesser degrees of relatedness to the class III receptors including CSF-1/Fms, PDGR, SLFR/Kit and Flt-3/Flk-2. Two members of this receptor class, Flt-1 and Flk-1, have been shown to represent high affinity receptors for vascularMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol FLT1
Antigen name VEGFR1 / Flt-1
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Clone FLT1/658
Reactivity Human
Host Mouse
Gene ID Unigene ID: 594454
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR