Anti-CD14 Mouse Monoclonal Antibody (CF568) [clone: LPSR/654]

Supplier: Biotium

BNC680654-100 BNC680654-500
75887-374EA 325.12 CAD
75887-374 75887-376
Anti-CD14 Mouse Monoclonal Antibody (CF568) [clone: LPSR/654]
Antibodies
This antibody recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: Clone LPSR/654
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR