Anti-FGF23 Mouse Monoclonal Antibody (CF594) [clone: FGF23/638]

Supplier: Biotium

BNC940638-100 BNC940638-500
75926-676EA 325.12 CAD
75926-676 75926-678
Anti-FGF23 Mouse Monoclonal Antibody (CF594) [clone: FGF23/638]
Antibodies
Fibroblast growth factor-1 (FGF-1), also designated acidic FGF, and fibroblast growth factor-2 (FGF-2), also designated basic FGF, are members of a family of growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuroectodermal origin. Additional members of the FGF family include the oncogenes FGF-3 (Int2) and FGF-4 (hst/Kaposi), FGF-5, FGF-6, FGF-7 (KGF), FGF-8 (AIGF), FGF-9 (GAF) and FGF-10 through FGF-23. Members of the FGF family share 30-55% amino acid sequence identity and similar gene structure, and are capable of transforming cultured cells whenMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol FGF23
Environmentally Preferable
Antigen name FGF23
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 594
Clone FGF23/638
Reactivity Human
Host Mouse
Gene ID Unigene ID: 287370
Isotype IgG1 kappa
ELISA Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR