Anti-CD2 Mouse Monoclonal Antibody (Biotin) [clone: LFA2/600]

Supplier: Biotium

BNCB0600-100 BNCB0600-500
75890-290EA 325.12 CAD
75890-290 75890-292
Anti-CD2 Mouse Monoclonal Antibody (Biotin) [clone: LFA2/600]
Antibodies
CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitating communication between helper T lymphocytes and antigen-presenting cells, as well as between cytolytic effectors and target cells.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: Clone LFA2/600
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Non-human primates, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR