Anti-MYC Mouse Monoclonal Antibody (CF555) [clone: 9E10.]

Supplier: Biotium

BNC550596-100 BNC550596-500
75921-022EA 325.12 CAD
75921-022 75922-154
Anti-MYC Mouse Monoclonal Antibody (CF555) [clone: 9E10.]
Antibodies
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. Its epitope spans aa 410-419 (EQKLISEEDL), which is a specific portion of an alpha helical region of human c-myc protein. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Overexpression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®555 is a bright orange-red fluorescent dye (Ex/Em 555/565 nm) that is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: Clone 9E10.
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR