Anti-NCL Mouse Monoclonal Antibody (CF640R) [clone: 364-5]
Supplier: Biotium
BNC400527-100 BNC400527-500
75966-444EA 325.12 CAD
75966-444 75966-446
Anti-NCL Mouse Monoclonal Antibody (CF640R) [clone: 364-5]
Antibodies
This antibody recognizes a protein of ~76 kDa, which is identified as Nucleolin (NCL). It is the major nucleolar phosphoprotein of growing eukaryotic cells. NCL is located mainly in dense fibrillar regions of the nucleolus. It is found associated with intranucleolar chromatin and pre-ribosomal particles. Human NCL gene consists of 14 exons with 13 introns and spans approximately 11kb. It induces chromatin decondensation by binding to histone H1. It is thought to play a role in pre-rRNA transcription and ribosome assembly.This MAb can be used to stain the nucleoli in cell or tissueMore Product Information