Anti-AFP Mouse Monoclonal Antibody (Biotin) [clone: MBS-12]

Supplier: Biotium

BNCB0521-100 BNCB0521-500
75883-862EA 325.12 CAD
75883-862 75883-864
Anti-AFP Mouse Monoclonal Antibody (Biotin) [clone: MBS-12]
Antibodies
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. The yolk sac and the liver produce AFP during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the AFP and albumin genes are present inMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol AFP AFP
Environmentally Preferable
Antigen name AFP AFP
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Conjugation Biotin Biotin
Clone MBS-12 MBS-12
Reactivity Human Human
Host Mouse Mouse
Gene ID Unigene ID: 518808 Unigene ID: 518808
Isotype IgG1 kappa IgG1 kappa
ImmunoChemistry Yes Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR