Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: ER505]

Supplier: Biotium

BNC680505-100 BNC680505-500
75925-060EA 325.12 CAD
75925-060 75925-062
Anti-ESR1 Mouse Monoclonal Antibody (CF568) [clone: ER505]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy. This MAb is excellent for staining of formalin-fixed, paraffin-embedded breast carcinomas.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone ER505
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR