Anti-KRT14 Mouse Monoclonal Antibody (CF568) [clone: LL002]

Supplier: Biotium

BNC680499-100 BNC680499-500
75916-288EA 325.12 CAD
75916-288 75916-290
Anti-KRT14 Mouse Monoclonal Antibody (CF568) [clone: LL002]
Antibodies
Cytokeratin 14 (CK14) belongs to the type I (or A or acidic) subfamily of low molecular weight keratins and exists in combination with keratin 5 (type II or B or basic). CK14 is found in basal cells of squamous epithelia, some glandular epithelia, myoepithelium, and mesothelial cells. Anti-CK14 is useful in differentiating squamous cell carcinomas from poorly differentiated epithelial tumors. Anti-CK14 is one of the specific basal markers for distinguishing between basal and non-basal subtypes of breast carcinomas. Anti-CK14 is also a good marker for differentiation of intraductal fromMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KRT14
Antigen name Cytokeratin 14
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 568
Clone LL002
Reactivity Human,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 6545380
Isotype IgG3
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR