Anti-PAX6 Mouse Monoclonal Antibody (CF488A) [clone: PAX6/498]

Supplier: Biotium

BNC880498-100 BNC880498-500
75968-220EA 325.12 CAD
75968-220 75968-222
Anti-PAX6 Mouse Monoclonal Antibody (CF488A) [clone: PAX6/498]
Antibodies
Pax genes contain paired domains with strong homology to genes in Drosophila, which are involved in programming early development. Lesions in the Pax-6 gene account for most cases of aniridia, a congenital malformation of the eye, chiefly characterized by iris hypoplasia, which can cause blindness. Pax-6 is involved in other anterior segment malformations besides aniridia, such as Peters anomaly, a major error in the embryonic development of the eye with corneal clouding with variable iridolenticulocorneal adhesions. The Pax-6 gene encodes a transcriptional regulator that recognizes targetMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol PAX6
Antigen name PAX6
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 488A
Clone PAX6/498
Reactivity Chicken,Human,Rat,Mouse
Host Mouse
Gene ID Unigene ID: 611376
Isotype IgG1 kappa
Western blot Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR