Anti-TOP1MT Mouse Monoclonal Antibody (CF640R) [clone: TOP1MT/488]

Supplier: Biotium

BNC400488-100 BNC400488-500
75977-364EA 325.12 CAD
75977-364 75977-366
Anti-TOP1MT Mouse Monoclonal Antibody (CF640R) [clone: TOP1MT/488]
Antibodies
DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA in eukaryotic cells by transiently breaking and rejoining DNA strands. Due to their roles in DNA replication, recombination, and transcription, DNA topoisomerases have been identified as targets of numerous anticancer drugs. Mitochondrial Topo I (DNA topoisomerase I, mitochondrial) is a 601 amino acid protein that primarily acts to relieve DNA strain that may occur during duplication of mitochondrial DNA. As a type IB topoisomerase, mitochondrial Topo I requires a divalent metal, either, calcium or magnesium, as well as an alkaline pH for optimal activity.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®640R is a far-red fluorescent dye (Ex/Em 642/662 nm) with excellent brightness, and the best photostabiity among spectrally-similar dyes.

Type: Primary
Antigen: TOP1MT
Clonality: Monoclonal
Clone: Clone TOP1MT/488
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR