Anti-ODC1 Mouse Monoclonal Antibody (CF647) [clone: ODC1/485]

Supplier: Biotium

BNC470485-500 BNC470485-100
75966-650EA 900.17 CAD
75966-650 75968-776
Anti-ODC1 Mouse Monoclonal Antibody (CF647) [clone: ODC1/485]
Antibodies
This antibody recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater thanMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ODC1
Antigen name ODC-1
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation CF® 647
Clone ODC1/485
Reactivity Human,Rat
Host Mouse
Gene ID Unigene ID: 467701
Isotype IgG1 kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR