Anti-ODC1 Mouse Monoclonal Antibody (CF405M) [clone: ODC1/485]

Supplier: Biotium

BNC050485-100 BNC050485-500
75966-640EA 325.12 CAD
75966-640 75966-642
Anti-ODC1 Mouse Monoclonal Antibody (CF405M) [clone: ODC1/485]
Antibodies
This antibody recognizes a 53 kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405M is a blue fluorescent dye (Ex/Em 408/452 nm) with superior brightness and photostability compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: ODC1
Clonality: Monoclonal
Clone: Clone ODC1/485
Conjugation: CF405M
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR