Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF568) [clone: L243]

Supplier: Biotium

BNC680458-100 BNC680458-500
75951-452EA 325.12 CAD
75951-452 75951-454
Anti-HLA-DRB1 Mouse Monoclonal Antibody (CF568) [clone: L243]
Antibodies
This MAb reacts with the HLA-DRB1 antigen, a member of MHC class II molecules. It does not cross react with HLA-DP and HLA-DQ. It binds a conformational epitope on HLA-DR, which depends on the correct folding of the α/β heterodimer. This MAb has been reported to block mixed lymphocyte reactions. The L243 antibody recognizes a different epitope than the LN3 monoclonal antibody, and these antibodies do not cross-block binding to each other's respective epitopes. HLA-DR is a heterodimeric cell surface glycoprotein comprised of a 36kD alpha (heavy) chain and a 28kD beta (light) chain. It is expressed on B-cells, activated T-cells, monocytes/macrophages, dendritic cells and other non-professional APCs. In conjunction with the CD3/TCR complex and CD4 molecules, HLA-DR is critical for efficient peptide presentation to CD4 T cells. It is an excellent histiocytic marker in paraffin sections producing intense staining. True histiocytic neoplasms are similarly positive. HLA-DR antigens also occur on a variety of epithelial cells and their corresponding neoplastic counterparts.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: HLA-DRB1
Clonality: Monoclonal
Clone: Clone L243
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Baboon, Chimpanzee, Dog, Human, Monkey, Squirrel
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR