Anti-Cytokeratins Mouse Monoclonal Antibody (CF647) [clone: KRT/457]

Supplier: Biotium

BNC470457-100 BNC470457-500
75916-056EA 325.12 CAD
75916-056 75918-268
Anti-Cytokeratins Mouse Monoclonal Antibody (CF647) [clone: KRT/457]
Antibodies
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, including 59 kDa (CK4); 58 kDa (CK5); 56 kDa (CK6); 52 kDa (CK8); 56.5 kDa (CK10); 53 kDa (CK13) and 45 kDa (CK18). This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®647 is a far-red fluorescent dye (Ex/Em 650/665 nm) with excellent brightness. It also is compatible with super-resolution imaging by STORM.

Type: Primary
Antigen: Multiple
Clonality: Monoclonal
Clone: Clone KRT/457
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Frog, Goat, Guinea Pig, Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR