Anti-CD47 Mouse Monoclonal Antibody (CF405S) [clone: B6H12.2]

Supplier: Biotium

BNC040437-100 BNC040437-500
75913-722EA 325.12 CAD
75913-722 75913-724
Anti-CD47 Mouse Monoclonal Antibody (CF405S) [clone: B6H12.2]
Antibodies
This antibody reacts with Ig domain of CD47 protein. It has been shown to inhibit polymorphonuclear neutrophil (PMN) transmigration across cell monolayers and matrix. CD47, originally named integrin-associated protein (IAP), is a 50 kDa protein containing five membrane-spanning sequences and a short cytoplasmic tail. CD47 plays a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins. It is important in memory formation and synaptic plasticity in the hippocampus. CD47 may play a role in membrane transport and/or integrin dependent signal transduction.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: Clone B6H12.2
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR