Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: 8D6]

Supplier: Biotium

BNC940424-100 BNC940424-500
75886-914EA 325.12 CAD
75886-914 75886-916
Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: 8D6]
Antibodies
This MAb recognizes extracellular epitope 2 within the N-terminal Ig domain of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: Clone 8D6
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR