Anti-ESR1 Mouse Monoclonal Antibody (CF594) [clone: ESR1/420]

Supplier: Biotium

BNC940420-100 BNC940420-500
75926-016EA 325.12 CAD
75926-016 75926-018
Anti-ESR1 Mouse Monoclonal Antibody (CF594) [clone: ESR1/420]
Antibodies
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa300-550. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields the brightest conjugates among spectrally similar dyes, and has excellent photostability.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: Clone ESR1/420
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR