Anti-TGM2 Mouse Monoclonal Antibody (CF405S) [clone: TGM2/419]

Supplier: Biotium

BNC040419-100 BNC040419-500
75976-372EA 325.12 CAD
75976-372 75976-374
Anti-TGM2 Mouse Monoclonal Antibody (CF405S) [clone: TGM2/419]
Antibodies
This antibody recognizes a 77-85 kDa protein, identified as cellular or tissue transglutaminase II (TGase II). Transglutaminases are enzymes that catalyze the crosslinking of proteins by epsilon-gamma glutamyl lysine isopeptide bonds. While the primary structure of transglutaminases is not conserved, they all have the same amino acid sequence at their active sites and their activity is calcium-dependent. The protein encoded by this gene acts as a monomer, is induced by retinoic acid, and appears to be involved in apoptosis. Finally, the encoded protein is the autoantigen implicated in celiac disease. The identification of transglutaminase as the main antigen of endomysium antibodies allows a new diagnostic approach to celiac disease (CD), a genetic, immunologically mediated small bowel enteropathy that causes malabsorption. TGase II is implicated in programmed cell death, signal transduction, drug-resistance, cell growth, endocytosis, insulin secretion, cell adhesion, cataract formation, and wound healing.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®405S is a blue fluorescent dye (Ex/Em 404/431 nm) with superior brightness compared to other blue dyes; it is also compatible with super-resolution imaging by SIM. Note: Conjugates of blue fluorescent dyes are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colors.

Type: Primary
Antigen: TGM2
Clonality: Monoclonal
Clone: Clone TGM2/419
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human, Monkey, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR