Anti-CHGA Mouse Monoclonal Antibody (CF594) [clone: CGA/414]
Supplier: Biotium
BNC940414-100 BNC940414-500
75909-734EA 325.12 CAD
75909-734 75909-736
Anti-CHGA Mouse Monoclonal Antibody (CF594) [clone: CGA/414]
Antibodies
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raiseMore Product Information