Anti-CHGA Mouse Monoclonal Antibody (CF488A) [clone: CHGA/413]
Supplier: Biotium
BNC880413-100 BNC880413-500
75909-778EA 325.12 CAD
75909-778 75909-780
Anti-CHGA Mouse Monoclonal Antibody (CF488A) [clone: CHGA/413]
Antibodies
Chromogranin A is present in neuroendocrine cells throughout the body, including the neuroendocrine cells of the large and small intestine, adrenal medulla and pancreatic islets. It is an excellent marker for carcinoid tumors, pheochromocytomas, paragangliomas, and other neuroendocrine tumors. Co-expression of chromogranin A and neuron specific enolase (NSE) is common in neuroendocrine neoplasms. Reportedly, co-expression of certain keratins and chromogranin indicates neuroendocrine lineage. The presence of strong anti-chromogranin staining and absence of anti-keratin staining should raiseMore Product Information