Anti-BCL10 Mouse Monoclonal Antibody [clone: BL10/411]

Supplier: Biotium

BNUB0411-100 BNUB0411-500
75884-476EA 380.65 CAD
75884-476 75884-478
Anti-BCL10 Mouse Monoclonal Antibody [clone: BL10/411]
Antibodies
BCL10, with an N-terminal caspase recruitment domain (CARD), is found in a number of apoptotic regulatory molecules. It was identified through its direct involvement in t(1;14) of mucosa-associated lymphoid tissue (MALT) lymphoma. Expression of BCL10 was shown to induce NFkB activation in a NIK-dependent pathway. This MAb labels subpopulations of normal B and T cells and is a useful tool for the sub-classification of lymphomas. In MALT lymphomas with the t(1;14) translocation, while 55% of MALT lymphomas lacking this translocation exhibited the same labeling pattern, although at a much lower level.

Type: Primary
Antigen: BCL10
Clonality: Monoclonal
Clone: Clone BL10/411
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR