Anti-UPK3A Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]

Supplier: Biotium

BNC940409-100 BNC940409-500
75981-008EA 325.12 CAD
75981-008 75981-010
Anti-UPK3A Rabbit Polyclonal Antibody (CF594) [clone: Rabbit PAb]
Antibodies
Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer.

CF® dyes are Biotium's next-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol UPK3A
Antigen name UPK3A
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation CF® 594
Clone Rabbit PAb
Reactivity Human
Host Rabbit
Gene ID Unigene ID: 632787
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Concentration 0.1 mg/mL
Storage buffer PBS/0.1% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR