Anti-UPK3A Rabbit Polyclonal Antibody (CF568) [clone: Rabbit PAb]

Supplier: Biotium

BNC680409-100 BNC680409-500
75981-004EA 325.12 CAD
75981-004 75981-006
Anti-UPK3A Rabbit Polyclonal Antibody (CF568) [clone: Rabbit PAb]
Antibodies
Uroplakin-3A (UPK3A) is a component of the asymmetric unit membrane (AUM). It is a highly specialized bio-membrane made by terminally differentiated urothelial cells. The protein may play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. UPK3A also contributes to the formation of urothelial glycocalyx, which may play an important role in preventing bacterial adherence through FimH bacterial protein binding leading to bladder infection. UPK3A has been shown as a helpful marker for the detection of bladder cancer.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: UPK3A
Clonality: Polyclonal
Clone: Clone Rabbit PAb
Conjugation: CF568
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR